Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008744.1 | NP_001008744.1 | 6 | 849 | |
NM_018319.3 | NP_060789.2 | 7 | 1072 | ||
XM_005267847.2 | XP_005267904.1 | 7 | 1746 | ||
XM_005267848.2 | XP_005267905.1 | 6 | 1090 | ||
XM_006720197.3 | XP_006720260.1 | 7 | 1313 | ||
XM_006720198.3 | XP_006720261.1 | 6 | 968 | ||
XM_006720199.1 | XP_006720262.1 | 6 | 454 | ||
XM_011536942.2 | XP_011535244.1 | 6 | 1489 | ||
XM_011536943.2 | XP_011535245.1 | 6 | 882 | ||
XM_011536944.1 | XP_011535246.1 | 7 | 1073 | ||
XM_017021439.1 | XP_016876928.1 | 7 | 980 | ||
XM_017021440.1 | XP_016876929.1 | 6 | 872 | ||
XM_017021441.1 | XP_016876930.1 | 6 | 1061 | ||
XM_017021446.1 | XP_016876935.1 | 5 | 504 | ||
GenBank mRNA | AF182002.1 | - | 5 | 758 | |
AK001952.1 | - | 6 | 813 | ||
AK093235.1 | - | 8 | 1833 | ||
AK315549.1 | - | 6 | 797 | ||
BC006083.1 | - | 7 | 1016 | ||
BC015474.1 | - | 7 | 1023 | 3900062 | |
BC107877.1 | - | 7 | 1076 | 6597752 |