Gene Transcripts
- Gene Symbol
- UBQLN4
- Entrez Gene ID
- 56893
- Gene Name
- ubiquilin 4
- Gene Aliases
- A1U, A1Up, C1orf6, CIP75, UBIN
- Chromosome Location
- Chr. 1: 156035294 - 156053825 on Build GRCh38
- UniGene ID
- Hs.283739
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304342.1 |
NP_001291271.1
|
6, 7 |
1199 |
|
NM_020131.4 |
NP_064516.2
|
6, 7 |
1259 |
|
XM_005245348.3 |
XP_005245405.1
|
6, 7 |
1164 |
|
GenBank mRNA |
AB209566.1 |
- |
6, 7 |
1132 |
|
AF113544.1 |
- |
3, 4 |
665 |
|
AF188240.1 |
- |
6, 7 |
1413 |
|
AK302895.1 |
- |
6, 7 |
1088 |
|
AK314413.1 |
- |
6, 7 |
1147 |
|
BC018403.2 |
- |
6, 7 |
1147 |
4214386 |
BC063841.1 |
- |
6, 7 |
1159 |
6183942 |
View Full Product Details