Gene Transcripts
- Gene Symbol
- CNN3
- Entrez Gene ID
- 1266
- Gene Name
- calponin 3
- Gene Aliases
- Chromosome Location
- Chr. 1: 94896949 - 94927223 on Build GRCh38
- UniGene ID
- Hs.483454
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286055.1 |
NP_001272984.1
|
2, 3 |
514 |
|
NM_001286056.1 |
NP_001272985.1
|
2, 3 |
304 |
|
NM_001839.4 |
NP_001830.1
|
2, 3 |
514 |
|
XM_017000245.1 |
XP_016855734.1
|
4, 5 |
2926 |
|
GenBank mRNA |
AK294141.1 |
- |
2, 3 |
265 |
|
AK298145.1 |
- |
2, 3 |
434 |
|
AK312370.1 |
- |
2, 3 |
401 |
|
AK316398.1 |
- |
2, 3 |
280 |
|
BC025372.2 |
- |
2, 3 |
398 |
3505668 |
BU149686.1 |
- |
2, 3 |
450 |
|
CR541848.1 |
- |
2, 3 |
185 |
|
DC422152.1 |
- |
2, 3 |
470 |
|
DR157009.1 |
- |
2, 3 |
437 |
|
S80562.1 |
- |
2, 3 |
268 |
|
View Full Product Details