Gene Transcripts

Gene Symbol
SCYL3
Entrez Gene ID
57147
Gene Name
SCY1 like pseudokinase 3
Gene Aliases
PACE-1, PACE1
Chromosome Location
Chr. 1: 169853074 - 169894267 on Build GRCh38
UniGene ID
Hs.435560
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020423.6 NP_065156.5 7 882
NM_181093.3 NP_851607.2 7 882
XM_006711465.1 XP_006711528.1 7 831
XM_011509801.1 XP_011508103.1 7 971
XM_011509802.1 XP_011508104.1 7 831
XM_011509803.1 XP_011508105.1 6 734
XM_017001862.1 XP_016857351.1 7 831
XM_017001863.1 XP_016857352.1 7 971
XM_017001864.1 XP_016857353.1 7 831
XM_017001865.1 XP_016857354.1 7 930
GenBank mRNA AF540957.1 - 6 712
AK292507.1 - 7 828
AK304481.1 - 2 474
AL117233.1 - 7 765
AY144493.1 - 6 712
BC014662.1 - 7 819 4852719
BX647352.1 - 7 971

View Full Product Details