Gene Transcripts

Gene Symbol
NIPAL3
Entrez Gene ID
57185
Gene Name
NIPA like domain containing 3
Gene Aliases
DJ462O23.2, NPAL3
Chromosome Location
Chr. 1: 24413360 - 24472983 on Build GRCh38
UniGene ID
Hs.523442
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322854.1 NP_001309783.1 5 919
NM_001322855.1 NP_001309784.1 6 1061
NM_001322856.1 NP_001309785.1 6 1018
NM_001322857.1 NP_001309786.1 5 839
NM_001322858.1 NP_001309787.1 6 936
NM_001322859.1 NP_001309788.1 3 545
NM_001322861.1 NP_001309790.1 2 195
NM_001322862.1 NP_001309791.1 6 896
NM_001322863.1 NP_001309792.1 5 868
NM_001322864.1 NP_001309793.1 4 436
NM_001322865.1 NP_001309794.1 5 786
NM_001322866.1 NP_001309795.1 5 759
NM_001322868.1 NP_001309797.1 5 786
NM_020448.4 NP_065181.1 5 781
NR_136506.1 5 786
NR_136508.1 5 786
XM_011541808.2 XP_011540110.1 4 696
XM_017001868.1 XP_016857357.1 4 431
XM_017001869.1 XP_016857358.1 5 766
XM_017001870.1 XP_016857359.1 6 1031
GenBank mRNA AK312433.1 - 5 735
BC001265.1 - 5 723 3452234
BC063583.1 - 5 711 4386115
BX640883.1 - 6 843

View Full Product Details