Gene Transcripts

Gene Symbol
NIPAL3
Entrez Gene ID
57185
Gene Name
NIPA like domain containing 3
Gene Aliases
DJ462O23.2, NPAL3
Chromosome Location
Chr. 1: 24413360 - 24472983 on Build GRCh38
UniGene ID
Hs.523442
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322854.1 NP_001309783.1 7 1155
NM_001322855.1 NP_001309784.1 8 1297
NM_001322856.1 NP_001309785.1 8 1254
NM_001322857.1 NP_001309786.1 7 1075
NM_001322858.1 NP_001309787.1 8 1172
NM_001322859.1 NP_001309788.1 5 781
NM_001322860.1 NP_001309789.1 4 721
NM_001322861.1 NP_001309790.1 4 431
NM_001322862.1 NP_001309791.1 8 1132
NM_001322863.1 NP_001309792.1 7 1104
NM_001322864.1 NP_001309793.1 6 672
NM_001322865.1 NP_001309794.1 7 1022
NM_001322866.1 NP_001309795.1 7 995
NM_001322868.1 NP_001309797.1 7 1022
NM_020448.4 NP_065181.1 7 1017
NR_136507.1 4 721
NR_136508.1 7 1022
XM_011541808.2 XP_011540110.1 6 932
XM_017001868.1 XP_016857357.1 6 667
XM_017001869.1 XP_016857358.1 7 1002
XM_017001870.1 XP_016857359.1 8 1267
GenBank mRNA AK312433.1 - 7 971
BC001265.1 - 7 959 3452234
BC063583.1 - 7 947 4386115
BX640883.1 - 8 1079
U90918.1 - 7 908

View Full Product Details