Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004369.3 | NP_004360.2 | 14 | 6326 | |
NM_057166.4 | NP_476507.3 | 11 | 4505 | ||
NM_057167.3 | NP_476508.2 | 13 | 5708 | ||
XM_005246065.1 | XP_005246122.1 | 13 | 5683 | ||
XM_005246066.1 | XP_005246123.1 | 11 | 4829 | ||
XM_006712253.1 | XP_006712316.1 | 14 | 5782 | ||
XM_011510574.1 | XP_011508876.1 | 14 | 6280 | ||
XM_017003303.1 | XP_016858792.1 | 19 | 6333 | ||
XM_017003304.1 | XP_016858793.1 | 16 | 4545 | ||
GenBank mRNA | AB209636.1 | - | 5 | 1617 | |
BC144595.1 | - | 11 | 4446 | 9053125 | |
BC150625.1 | - | 11 | 4446 | 9056995 | |
BC171790.1 | - | 11 | 4446 | 9054444 | |
X52022.1 | - | 14 | 6296 |