Gene Transcripts

Gene Symbol
WDR19
Entrez Gene ID
57728
Gene Name
WD repeat domain 19
Gene Aliases
ATD5, CED4, DYF-2, IFT144, NPHP13, ORF26, Oseg6, PWDMP, SRTD5
Chromosome Location
Chr. 4: 39182404 - 39285810 on Build GRCh38
UniGene ID
Hs.438482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317924.1 NP_001304853.1 24 2832
NM_025132.3 NP_079408.3 25 2948
XM_011513724.1 XP_011512026.1 25 2870
XM_011513725.2 XP_011512027.1 24 2825
XM_011513726.2 XP_011512028.1 24 2733
XM_011513727.1 XP_011512029.1 23 2653
XM_017008501.1 XP_016863990.1 23 2641
GenBank mRNA AB046858.1 - 15 1792
AK294730.1 - 24 2707
AK296112.1 - 9 922
AK304161.1 - 24 2641
AY029257.1 - 25 2948

View Full Product Details