Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127173.1 | NP_001120645.1 | 5 | 786 | |
NM_021189.3 | NP_067012.1 | 6 | 925 | ||
GenBank mRNA | AF062733.2 | - | 6 | 882 | |
AF363367.1 | - | 5 | 784 | ||
AK001560.1 | - | 2 | 184 | ||
AK289541.1 | - | 6 | 925 | ||
AY046418.1 | - | 5 | 689 | ||
AY358332.1 | - | 5 | 790 | ||
BC033819.1 | - | 5 | 734 | 5199627 |