Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005260476.2 | XP_005260533.1 | 5 | 1376 | |
XM_006723841.2 | XP_006723904.1 | 3 | 1022 | ||
XM_011528952.2 | XP_011527254.1 | 5 | 1437 | ||
XM_011528953.2 | XP_011527255.1 | 4 | 1317 | ||
GenBank mRNA | AJ297792.1 | - | 5 | 1384 | |
AL137597.1 | - | 1 | 204 | ||
AY422170.1 | - | 5 | 1393 | ||
BC035639.1 | - | 5 | 1282 | 5547220 |