Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001866.2 | NP_001857.1 | 2 | 203 | |
GenBank mRNA | AK311879.1 | - | 2 | 185 | |
BC018386.1 | - | 2 | 186 | 3861730 | |
BT009767.1 | - | 2 | 113 | ||
CR450332.1 | - | 2 | 113 | ||
CR542124.1 | - | 2 | 113 | ||
Z14244.1 | - | 2 | 215 |