Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022062.2 | NP_071345.2 | 7 | 787 | |
XM_005271642.2 | XP_005271699.1 | 6 | 676 | ||
XM_005271643.2 | XP_005271700.1 | 4 | 493 | ||
XM_006718894.2 | XP_006718957.1 | 6 | 563 | ||
XM_011542944.2 | XP_011541246.1 | 7 | 872 | ||
XM_011542945.2 | XP_011541247.1 | 8 | 832 | ||
XM_011542946.1 | XP_011541248.1 | 7 | 755 | ||
XM_011542947.2 | XP_011541249.1 | 5 | 441 | ||
XM_017018110.1 | XP_016873599.1 | 7 | 3622 | ||
XM_017018111.1 | XP_016873600.1 | 5 | 482 | ||
XM_017018112.1 | XP_016873601.1 | 4 | 493 | ||
GenBank mRNA | AB065001.1 | - | 4 | 465 | |
AJ417081.1 | - | 4 | 437 | ||
AK023136.1 | - | 4 | 465 | ||
AK295857.1 | - | 6 | 654 | ||
AK298999.1 | - | 6 | 556 | ||
AK316001.1 | - | 6 | 587 | ||
AK316268.1 | - | 5 | 575 | ||
AL512682.1 | - | 2 | 110 | ||
BC045626.1 | - | 7 | 744 | 5261106 | |
BX648615.1 | - | 5 | 564 |