Gene Transcripts

Gene Symbol
SNX16
Entrez Gene ID
64089
Gene Name
sorting nexin 16
Gene Aliases
Chromosome Location
Chr. 8: 81799583 - 81842326 on Build GRCh38
UniGene ID
Hs.492121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022133.3 NP_071416.2 6 1166
NM_152836.2 NP_690049.1 5 920
NM_152837.2 NP_690050.1 4 833
XM_005251282.4 XP_005251339.1 6 873
XM_005251283.2 XP_005251340.1 7 1010
XM_011517574.2 XP_011515876.1 4 263
GenBank mRNA AF305779.1 - 5 771
AK290903.1 - 5 836
AL833763.1 - 4 740
BC033630.1 - 5 828 5224216

View Full Product Details