Gene Transcripts

Gene Symbol
POPDC2
Entrez Gene ID
64091
Gene Name
popeye domain containing 2
Gene Aliases
POP2
Chromosome Location
Chr. 3: 119642052 - 119660590 on Build GRCh38
UniGene ID
Hs.534383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308333.1 NP_001295262.1 3 883
NM_022135.3 NP_071418.2 3 883
XM_005247698.3 XP_005247755.1 3 883

View Full Product Details