Gene Transcripts

Gene Symbol
GORASP1
Entrez Gene ID
64689
Gene Name
golgi reassembly stacking protein 1
Gene Aliases
GOLPH5, GRASP65, P65
Chromosome Location
Chr. 3: 39096599 - 39108363 on Build GRCh38
UniGene ID
Hs.721946
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278789.1 NP_001265718.1 3 534
NM_031899.3 NP_114105.1 5 1282
NR_103866.1 5 819
NR_103867.1 5 819
XM_006713301.3 XP_006713364.1 5 1382
XM_011534020.2 XP_011532322.1 5 1382
XM_017007050.1 XP_016862539.1 5 558
GenBank mRNA AJ409349.1 - 5 558
AK001574.1 - 5 557
AK091168.1 - 5 746
AK124755.1 - 5 1282
AK300271.1 - 4 636
AK303847.1 - 3 274
BC008928.2 - 5 540
BC075854.1 - 5 568 5503263
BC103826.1 - 5 500 40001584
BC103827.1 - 5 500 40001588
DB031221.1 - 5 557

View Full Product Details