Gene Transcripts

Gene Symbol
LPIN3
Entrez Gene ID
64900
Gene Name
lipin 3
Gene Aliases
LIPN3L, SMP2
Chromosome Location
Chr. 20: 41340779 - 41360585 on Build GRCh38
UniGene ID
Hs.25897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301860.1 NP_001288789.1 4 606
NR_126051.1 4 606
XM_005260516.2 XP_005260573.1 4 604
XM_006723863.2 XP_006723926.1 4 604
XM_011528996.1 XP_011527298.1 4 604
XM_011528997.2 XP_011527299.1 5 810
XM_011528998.2 XP_011527300.1 4 627
XM_011528999.2 XP_011527301.1 5 757
XM_011529000.1 XP_011527302.1 4 604
XM_011529001.1 XP_011527303.1 4 604
XM_011529002.2 XP_011527304.1 3 406
XM_011529003.1 XP_011527305.1 4 604
XM_011529004.1 XP_011527306.1 4 604
XM_011529005.2 XP_011527307.1 4 604
XM_011529006.2 XP_011527308.1 4 604
XM_017028020.1 XP_016883509.1 4 604
GenBank mRNA BC140806.1 - 4 487
BX647082.1 - 4 606

View Full Product Details