Gene Transcripts

Gene Symbol
MAPK14
Entrez Gene ID
1432
Gene Name
mitogen-activated protein kinase 14
Gene Aliases
CSBP, CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA
Chromosome Location
Chr. 6: 36027635 - 36122964 on Build GRCh38
UniGene ID
Hs.485233
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001315.2 NP_001306.1 2 703
NM_139012.2 NP_620581.1 2 703
NM_139013.2 NP_620582.1 2 703
NM_139014.2 NP_620583.1 2 703
XM_006714998.3 XP_006715061.1 2 178
XM_011514310.2 XP_011512612.2 2 971
XM_017010299.1 XP_016865788.1 2 745
XM_017010300.1 XP_016865789.1 2 745
XM_017010301.1 XP_016865790.1 2 745
XM_017010302.1 XP_016865791.1 2 745
XM_017010303.1 XP_016865792.1 2 745
XM_017010304.1 XP_016865793.1 2 745
GenBank mRNA AB074150.1 - 2 222
AF100544.1 - 2 237
AK291709.1 - 2 598
AK303414.1 - 3 267
BC000092.1 - 2 560 3505234
BC031574.1 - 2 559 5181064
BT006933.1 - 2 222
CR536505.1 - 2 222
FJ032367.1 - 2 222
FJ032368.1 - 2 222
L35263.1 - 2 584
L35264.1 - 2 430

View Full Product Details