Gene Transcripts
- Gene Symbol
- RHBDF2
- Entrez Gene ID
- 79651
- Gene Name
- rhomboid 5 homolog 2
- Gene Aliases
- RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2
- Chromosome Location
- Chr. 17: 76470893 - 76501440 on Build GRCh38
- UniGene ID
- Hs.464157
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005498.3 |
NP_001005498.2
|
4, 5 |
691 |
|
NM_024599.5 |
NP_078875.4
|
5, 6 |
853 |
|
XM_005257669.3 |
XP_005257726.1
|
5, 6 |
1214 |
|
XM_005257670.1 |
XP_005257727.1
|
5, 6 |
723 |
|
XM_011525249.2 |
XP_011523551.1
|
5, 6 |
2170 |
|
XM_011525250.2 |
XP_011523552.1
|
6, 7 |
880 |
|
XM_011525251.2 |
XP_011523553.1
|
5, 6 |
697 |
|
XM_017025079.1 |
XP_016880568.1
|
5, 6 |
1127 |
|
GenBank mRNA |
AK074034.1 |
- |
3, 4 |
1158 |
|
AK292135.1 |
- |
5, 6 |
846 |
|
AK292166.1 |
- |
4, 5 |
646 |
|
AK303434.1 |
- |
3, 4 |
507 |
|
AY500247.1 |
- |
4, 5 |
542 |
|
BC016034.1 |
- |
5, 6 |
833 |
3910543 |
DC348660.1 |
- |
3, 4 |
443 |
|
View Full Product Details