Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205315.1 | NP_001192244.1 | 4 | 1084 | |
NM_024636.3 | NP_078912.2 | 3 | 985 | ||
GenBank mRNA | AF423422.1 | - | 4 | 967 | |
AF423423.1 | - | 4 | 967 | ||
AK026806.1 | - | 3 | 965 | ||
AK307644.1 | - | 3 | 963 | ||
AL832955.1 | - | 1 | 76 |