Gene Transcripts

Gene Symbol
ANKRD55
Entrez Gene ID
79722
Gene Name
ankyrin repeat domain 55
Gene Aliases
Chromosome Location
Chr. 5: 56099680 - 56233359 on Build GRCh38
UniGene ID
Hs.436214
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024669.2 NP_078945.2 10 1565
XM_011543646.1 XP_011541948.1 4 879
XM_017009852.1 XP_016865341.1 10 1477
XM_017009853.1 XP_016865342.1 9 1449
XM_017009854.1 XP_016865343.1 7 2799
GenBank mRNA AK021857.1 - 2 618
AK123389.1 - 10 1565
BC106914.2 - 2 785 40032109
BC106915.2 - 2 785 40032110

View Full Product Details