Gene Transcripts

Gene Symbol
TBC1D17
Entrez Gene ID
79735
Gene Name
TBC1 domain family member 17
Gene Aliases
Chromosome Location
Chr. 19: 49877425 - 49888750 on Build GRCh38
UniGene ID
Hs.631587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168222.1 NP_001161694.1 9 1269
NM_024682.2 NP_078958.2 10 1368
XM_011527317.2 XP_011525619.1 10 1280
GenBank mRNA AB449903.1 - 10 1069
AK022230.1 - 10 1127
AK090606.1 - 9 1069
AK300007.1 - 9 1028
AY129013.1 - 8 1815
BC003516.1 - 10 1221 3050014

View Full Product Details