Gene Transcripts
- Gene Symbol
- CLIP4
- Entrez Gene ID
- 79745
- Gene Name
- CAP-Gly domain containing linker protein family member 4
- Gene Aliases
- RSNL2
- Chromosome Location
- Chr. 2: 29097676 - 29183813 on Build GRCh38
- UniGene ID
- Hs.122927
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287527.1 |
NP_001274456.1
|
12, 13 |
1790 |
|
NM_001287528.1 |
NP_001274457.1
|
12, 13 |
1826 |
|
NM_024692.5 |
NP_078968.3
|
12, 13 |
1794 |
|
NR_109844.1 |
|
11, 12 |
1787 |
|
XM_005264562.2 |
XP_005264619.1
|
12, 13 |
1821 |
|
XM_005264563.1 |
XP_005264620.1
|
12, 13 |
1826 |
|
XM_006712104.2 |
XP_006712167.1
|
12, 13 |
1866 |
|
XM_006712105.3 |
XP_006712168.1
|
12, 13 |
1708 |
|
XM_011533109.1 |
XP_011531411.1
|
12, 13 |
1574 |
|
XM_011533110.1 |
XP_011531412.1
|
11, 12 |
1627 |
|
XM_017004958.1 |
XP_016860447.1
|
12, 13 |
1653 |
|
XM_017004959.1 |
XP_016860448.1
|
12, 13 |
1708 |
|
GenBank mRNA |
AB209042.1 |
- |
12, 13 |
3460 |
|
AK024722.1 |
- |
9, 10 |
1237 |
|
AK057267.1 |
- |
11, 12 |
1788 |
|
AK225074.1 |
- |
9, 10 |
1237 |
|
AK302325.1 |
- |
12, 13 |
1776 |
|
AK304384.1 |
- |
12, 13 |
1554 |
|
AL834443.1 |
- |
12, 13 |
1768 |
|
BC013862.1 |
- |
12, 13 |
1774 |
|
BC126167.1 |
- |
11, 12 |
1564 |
8991883 |
BC136331.1 |
- |
11, 12 |
1564 |
9020318 |
View Full Product Details