Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024769.3 | NP_079045.1 | 4 | 775 | |
XM_011542998.2 | XP_011541300.1 | 4 | 743 | ||
GenBank mRNA | AK026068.1 | - | 4 | 751 | |
AK304843.1 | - | 4, 5 | 722 | ||
AY326422.1 | - | 4 | 599 | ||
AY358340.1 | - | 4 | 599 | ||
BC009371.2 | - | 4 | 421 | 4122648 |