Gene Transcripts

Gene Symbol
KDM8
Entrez Gene ID
79831
Gene Name
lysine demethylase 8
Gene Aliases
JMJD5
Chromosome Location
Chr. 16: 27203477 - 27222305 on Build GRCh38
UniGene ID
Hs.145717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145348.1 NP_001138820.1 2 491
NM_024773.2 NP_079049.2 2 441
XM_017023676.1 XP_016879165.1 2 441
XM_017023677.1 XP_016879166.1 2 450
XM_017023678.1 XP_016879167.1 2 450
XM_017023679.1 XP_016879168.1 2 450
XM_017023680.1 XP_016879169.1 1 200
GenBank mRNA AK023860.1 - 2 432
AK225066.1 - 2 398
AK297166.1 - 2 491
AY345239.1 - 1 259
BC027911.1 - 2 420 5207043
DA497245.1 - 2 442
KU178785.1 - 1 259
KU178786.1 - 1 259
KU178787.1 - 1 259

View Full Product Details