Gene Transcripts

Gene Symbol
SLC35G2
Entrez Gene ID
80723
Gene Name
solute carrier family 35 member G2
Gene Aliases
TMEM22
Chromosome Location
Chr. 3: 136819019 - 136855892 on Build GRCh38
UniGene ID
Hs.477692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097599.1 NP_001091068.1 2 1539
NM_001097600.1 NP_001091069.1 2 1390
NM_025246.2 NP_079522.2 2 1804
XM_006713773.3 XP_006713836.1 2 3956
XM_011513214.2 XP_011511516.1 2 2143
XM_017007289.1 XP_016862778.1 2 1267
XM_017007290.1 XP_016862779.1 2 1466
XM_017007291.1 XP_016862780.1 2 1272
GenBank mRNA AK314079.1 - 2 1699
AL136661.1 - 2 1685
BC000111.1 - 2 1280 3508204
BC022557.1 - 2 1325 4793991
BC050423.1 - 2 1804

View Full Product Details