Gene Transcripts
- Gene Symbol
- EGFL8
- Entrez Gene ID
- 80864
- Gene Name
- EGF like domain multiple 8
- Gene Aliases
- C6orf8, NG3
- Chromosome Location
- Chr. 6: 32164605 - 32168285 on Build GRCh38
- UniGene ID
- Hs.731888
- Species
- Homo sapiens
- Gene Symbol
- PPT2-EGFL8
- Entrez Gene ID
- 100532746
- Gene Name
- PPT2-EGFL8 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 6: 32153999 - 32168285 on Build GRCh38
- UniGene ID
- Hs.731888
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037861.1 |
|
14 |
2164 |
|
GenBank mRNA |
AF020544.1 |
- |
14 |
2318 |
|
View Full Product Details