Gene Transcripts

Gene Symbol
OR4A16
Entrez Gene ID
81327
Gene Name
olfactory receptor family 4 subfamily A member 16
Gene Aliases
OR11-117, OR4A16Q
Chromosome Location
Chr. 11: 55343201 - 55344187 on Build GRCh38
UniGene ID
Hs.554530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005274.1 NP_001005274.1 1 133
GenBank mRNA BC140748.1 - 1 183

View Full Product Details