Gene Transcripts
- Gene Symbol
- SLC38A1
- Entrez Gene ID
- 81539
- Gene Name
- solute carrier family 38 member 1
- Gene Aliases
- ATA1, NAT2, SAT1, SNAT1
- Chromosome Location
- Chr. 12: 46183055 - 46269425 on Build GRCh38
- UniGene ID
- Hs.533770
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001077484.1 |
NP_001070952.1
|
15, 16 |
2041 |
|
NM_001278387.1 |
NP_001265316.1
|
16, 17 |
2503 |
|
NM_001278388.1 |
NP_001265317.1
|
14, 15 |
2265 |
|
NM_001278389.1 |
NP_001265318.1
|
15, 16 |
1530 |
|
NM_001278390.1 |
NP_001265319.1
|
15, 16 |
2243 |
|
NM_030674.3 |
NP_109599.3
|
15, 16 |
2380 |
|
XM_011538784.2 |
XP_011537086.1
|
15, 16 |
1765 |
|
XM_011538786.2 |
XP_011537088.1
|
15, 16 |
1765 |
|
XM_011538787.2 |
XP_011537089.1
|
15, 16 |
1765 |
|
XM_017019990.1 |
XP_016875479.1
|
15, 16 |
1995 |
|
GenBank mRNA |
AF247166.1 |
- |
15, 16 |
1726 |
|
AF271070.1 |
- |
15, 16 |
1624 |
|
AK027825.1 |
- |
15, 16 |
1530 |
|
AK074758.1 |
- |
8, 9 |
797 |
|
AK074797.1 |
- |
12, 13 |
1099 |
|
AK074851.1 |
- |
13, 14 |
1242 |
|
AK074949.1 |
- |
13, 14 |
1288 |
|
AK225639.1 |
- |
15, 16 |
1618 |
|
AK225778.1 |
- |
15, 16 |
1640 |
|
AK309410.1 |
- |
15, 16 |
1957 |
|
AK313731.1 |
- |
13, 14 |
1331 |
|
BC010620.1 |
- |
15, 16 |
1587 |
3871101 |
BX647547.1 |
- |
16, 17 |
2075 |
|
BX648495.1 |
- |
15, 16 |
1968 |
|
BX648676.1 |
- |
14, 15 |
1835 |
|
View Full Product Details