Gene Transcripts

Gene Symbol
AGMAT
Entrez Gene ID
79814
Gene Name
agmatinase
Gene Aliases
Chromosome Location
Chr. 1: 15571699 - 15585110 on Build GRCh38
UniGene ID
Hs.461532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024758.4 NP_079034.3 7 1350
GenBank mRNA AK130034.1 - 7 1146
AY057097.1 - 7 1250

View Full Product Details