Gene Transcripts
- Gene Symbol
- CYP2A6
- Entrez Gene ID
- 1548
- Gene Name
- cytochrome P450 family 2 subfamily A member 6
- Gene Aliases
- CPA6, CYP2A, CYP2A3, CYPIIA6, P450C2A, P450PB
- Chromosome Location
- Chr. 19: 40843538 - 40850447 on Build GRCh38
- UniGene ID
- Hs.250615
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000762.5 |
NP_000753.3
|
1, 2 |
199 |
|
GenBank mRNA |
AF182275.1 |
- |
1, 2 |
187 |
|
AK312964.1 |
- |
1, 2 |
199 |
|
BC096253.3 |
- |
1, 2 |
187 |
40006061 |
BC096254.1 |
- |
1, 2 |
187 |
40006062 |
BC096255.1 |
- |
1, 2 |
187 |
40006064 |
BC096256.1 |
- |
1, 2 |
187 |
40006068 |
DA641580.1 |
- |
1, 2 |
199 |
|
M33318.1 |
- |
1, 2 |
187 |
|
X13897.1 |
- |
1, 2 |
173 |
|
X13929.1 |
- |
1, 2 |
41 |
|
X13930.1 |
- |
1, 2 |
187 |
|
- Gene Symbol
- CYP2A13
- Entrez Gene ID
- 1553
- Gene Name
- cytochrome P450 family 2 subfamily A member 13
- Gene Aliases
- CPAD, CYP2A, CYPIIA13
- Chromosome Location
- Chr. 19: 41088451 - 41096195 on Build GRCh38
- UniGene ID
- Hs.567252
- Species
- Homo sapiens
View Full Product Details