Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286777.1 | NP_001273706.1 | 12 | 1262 | |
NM_001286778.1 | NP_001273707.1 | 13 | 1378 | ||
NM_031461.5 | NP_113649.1 | 14 | 1890 | ||
GenBank mRNA | AF142573.1 | - | 14 | 1486 | |
AF329197.1 | - | 13 | 1465 | ||
AK304034.1 | - | 13 | 1366 | ||
AK304350.1 | - | 12 | 1243 | ||
AK314048.1 | - | 14 | 1646 | ||
AL834301.1 | - | 14 | 1622 | ||
AY358564.1 | - | 14 | 1848 | ||
BC020514.1 | - | 14 | 1755 | 3856615 |