Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031471.5 | NP_113659.3 | 10 | 1295 | |
NM_178443.2 | NP_848537.1 | 10 | 1307 | ||
XM_011545294.2 | XP_011543596.1 | 10 | 1319 | ||
XM_011545295.2 | XP_011543597.1 | 9 | 1152 | ||
XM_017018398.1 | XP_016873887.1 | 10 | 1303 | ||
XM_017018399.1 | XP_016873888.1 | 9 | 1155 | ||
GenBank mRNA | AK093719.1 | - | 10 | 1240 | |
AY093951.1 | - | 10 | 1241 | ||
AY093952.1 | - | 10 | 1253 | ||
BC004347.1 | - | 10 | 1239 | 3634032 | |
BC013366.2 | - | 10 | 1239 | 3677274 | |
BC015584.2 | - | 10 | 1234 | 4637808 |