Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031949.4 | NP_114155.4 | 3 | 1208 | |
XM_006715572.3 | XP_006715635.1 | 2 | 1003 | ||
GenBank mRNA | AK093039.1 | - | 3 | 1208 | |
AK098527.1 | - | 1 | 649 | ||
AK225984.1 | - | 3 | 1208 | ||
AK301755.1 | - | 4 | 1065 | ||
AK314450.1 | - | 3 | 1185 | ||
BC030650.1 | - | 3 | 1198 | 5269216 | |
BC047411.1 | - | 3 | 1186 | 5296230 |