Gene Transcripts

Gene Symbol
HDAC10
Entrez Gene ID
83933
Gene Name
histone deacetylase 10
Gene Aliases
HD10
Chromosome Location
Chr. 22: 50245183 - 50251405 on Build GRCh38
UniGene ID
Hs.26593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159286.1 NP_001152758.1 3, 4 663
NM_032019.5 NP_114408.3 3, 4 663
GenBank mRNA AF393962.1 - 3, 4 290
AF407272.1 - 3, 4 290
AF407273.1 - 3, 4 290
AF426160.1 - 3, 4 643
AY450395.1 - 3, 4 290
BC032049.1 - 3, 4 476
BC125083.1 - 3, 4 337 40117772
BI916349.1 - 3, 4 535
CR456465.1 - 3, 4 346

View Full Product Details