Gene Transcripts
- Gene Symbol
- NBPF3
- Entrez Gene ID
- 84224
- Gene Name
- neuroblastoma breakpoint family member 3
- Gene Aliases
- AE2
- Chromosome Location
- Chr. 1: 21436775 - 21484900 on Build GRCh38
- UniGene ID
- Hs.325422
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256416.2 |
NP_001243345.1
|
3, 4 |
736 |
|
NM_032264.4 |
NP_115640.1
|
3, 4 |
736 |
|
NR_046176.2 |
|
6, 7 |
1265 |
|
XM_006710957.1 |
XP_006711020.1
|
3, 4 |
705 |
|
XM_006710958.1 |
XP_006711021.1
|
6, 7 |
1050 |
|
XM_006710959.2 |
XP_006711022.1
|
4, 5 |
834 |
|
XM_011542276.1 |
XP_011540578.1
|
7, 8 |
1307 |
|
XM_011542277.1 |
XP_011540579.1
|
7, 8 |
1292 |
|
XM_011542278.1 |
XP_011540580.1
|
7, 8 |
1121 |
|
XM_011542281.1 |
XP_011540583.1
|
4, 5 |
657 |
|
XM_017002498.1 |
XP_016857987.1
|
7, 8 |
1106 |
|
XM_017002499.1 |
XP_016857988.1
|
2, 3 |
372 |
|
XM_017002500.1 |
XP_016857989.1
|
6, 7 |
1236 |
|
XM_017002501.1 |
XP_016857990.1
|
7, 8 |
1121 |
|
XM_017002502.1 |
XP_016857991.1
|
6, 7 |
1050 |
|
GenBank mRNA |
AK095602.1 |
- |
6, 7 |
1218 |
|
AK292580.1 |
- |
3, 4 |
601 |
|
AK314597.1 |
- |
3, 4 |
664 |
|
AL136890.1 |
- |
3, 4 |
513 |
|
AY598326.1 |
- |
2, 3 |
338 |
|
AY894564.1 |
- |
2, 3 |
242 |
|
View Full Product Details