Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000574.4 | NP_000565.1 | 2 | 552 | |
NM_001114752.2 | NP_001108224.1 | 2 | 552 | ||
NM_001300902.1 | NP_001287831.1 | 2 | 552 | ||
NM_001300903.1 | NP_001287832.1 | 2 | 552 | ||
NM_001300904.1 | NP_001287833.1 | 2 | 552 | ||
NR_125349.1 | 2 | 552 | |||
XM_017000467.1 | XP_016855956.1 | 2 | 386 | ||
GenBank mRNA | AB240566.1 | - | 2 | 276 | |
AB240567.1 | - | 2 | 276 | ||
AB240568.1 | - | 2 | 276 | ||
AB240569.1 | - | 2 | 291 | ||
AB240570.1 | - | 2 | 291 | ||
AF052110.1 | - | 2 | 492 | ||
AK298549.1 | - | 2 | 340 | ||
AK300621.1 | - | 2 | 344 | ||
AK312448.1 | - | 2 | 347 | ||
BC001288.1 | - | 2 | 337 | 3460621 | |
BI545505.1 | - | 2 | 521 | ||
BT007159.1 | - | 2 | 258 | ||
M30142.1 | - | 2 | 323 | ||
M31516.1 | - | 2 | 323 | ||
U88576.1 | - | 1 | 156 |