Gene Transcripts

Gene Symbol
MCHR2
Entrez Gene ID
84539
Gene Name
melanin concentrating hormone receptor 2
Gene Aliases
GPR145, GPRv17, MCH-2R, MCH-R2, MCH2, MCH2R, MCHR-2, SLT
Chromosome Location
Chr. 6: 99919910 - 99994238 on Build GRCh38
UniGene ID
Hs.591342
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040179.1 NP_001035269.1 2 475
NM_032503.2 NP_115892.2 2 251
GenBank mRNA AB058849.1 - 1 160
AB060151.1 - 1 160
AF347063.1 - 2 271
AF399937.1 - 1 160
AK123634.1 - 2 475
AY029596.1 - 1 160
AY562946.1 - 1 160
BC095515.1 - 1 187 6971912

View Full Product Details