Gene Transcripts

Gene Symbol
RETNLB
Entrez Gene ID
84666
Gene Name
resistin like beta
Gene Aliases
FIZZ1, FIZZ2, HXCP2, RELM-beta, RELMb, RELMbeta, XCP2
Chromosome Location
Chr. 3: 108755639 - 108757283 on Build GRCh38
UniGene ID
Hs.307047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032579.2 NP_115968.1 1, 2 235
GenBank mRNA AF290873.1 - 1, 2 237
AF323084.1 - 1, 2 244
AM050721.1 - 1, 2 139
AY358655.1 - 1, 2 236
BC069318.1 - 1, 2 208 7262095
BC113502.1 - 1, 2 169 8322554
BC113528.1 - 1, 2 169 8322580

View Full Product Details