Gene Transcripts
- Gene Symbol
- FAM71F1
- Entrez Gene ID
- 84691
- Gene Name
- family with sequence similarity 71 member F1
- Gene Aliases
- FAM137A, NYD-SP18
- Chromosome Location
- Chr. 7: 128709061 - 128731743 on Build GRCh38
- UniGene ID
- Hs.732440
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282788.1 |
NP_001269717.1
|
3, 4 |
787 |
|
NM_001282789.1 |
NP_001269718.1
|
4, 5 |
734 |
|
NM_032599.3 |
NP_115988.1
|
3, 4 |
787 |
|
NR_104242.1 |
|
3, 4 |
787 |
|
NR_104243.1 |
|
4, 5 |
676 |
|
XM_017012743.1 |
XP_016868232.1
|
3, 4 |
830 |
|
GenBank mRNA |
AF367470.1 |
- |
3, 4 |
740 |
|
AK093161.1 |
- |
4, 5 |
731 |
|
AK302221.1 |
- |
4, 5 |
575 |
|
AK310308.1 |
- |
3, 4 |
734 |
|
AK310426.1 |
- |
4, 5 |
643 |
|
AK313608.1 |
- |
3, 4 |
738 |
|
BC037248.1 |
- |
3, 4 |
726 |
5165912 |
View Full Product Details