Gene Transcripts

Gene Symbol
CRACR2A
Entrez Gene ID
84766
Gene Name
calcium release activated channel regulator 2A
Gene Aliases
EFCAB4B
Chromosome Location
Chr. 12: 3613366 - 3753216 on Build GRCh38
UniGene ID
Hs.504534
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144958.1 NP_001138430.1 8 1304
NM_032680.3 NP_116069.1 8 1304
XM_006719021.3 XP_006719084.1 8 1304
XM_011521034.2 XP_011519336.1 7 1051
XM_011521036.2 XP_011519338.1 6 811
XM_011521037.2 XP_011519339.1 2 149
XM_011521038.2 XP_011519340.1 2 146
GenBank mRNA AK091124.1 - 2 140
AK304017.1 - 8 1206
BC004524.2 - 8 1201
BC150643.1 - 7 935 9057013

View Full Product Details