Gene Transcripts

Gene Symbol
AGBL4
Entrez Gene ID
84871
Gene Name
ATP/GTP binding protein like 4
Gene Aliases
CCP6
Chromosome Location
Chr. 1: 48520157 - 50024011 on Build GRCh38
UniGene ID
Hs.551844
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323573.1 NP_001310502.1 9, 10 1155
NM_001323574.1 NP_001310503.1 9, 10 1155
NM_001323575.1 NP_001310504.1 9, 10 1119
NM_032785.3 NP_116174.3 9, 10 1119
NR_136623.1 9, 10 1118
XM_011542308.2 XP_011540610.1 10, 11 1214
XM_017002595.1 XP_016858084.1 9, 10 1176
XM_017002596.1 XP_016858085.1 9, 10 1176
XM_017002597.1 XP_016858086.1 9, 10 1176
GenBank mRNA AK027348.1 - 5, 6 498
AK094826.1 - 9, 10 1078
AK294394.1 - 9, 10 1114
AK294633.1 - 9, 10 1118
BC126383.1 - 5, 6 431 8992099
BC144171.1 - 4, 5 341

View Full Product Details