Gene Transcripts

Gene Symbol
MFSD2A
Entrez Gene ID
84879
Gene Name
major facilitator superfamily domain containing 2A
Gene Aliases
MCPH15, MFSD2, NLS1
Chromosome Location
Chr. 1: 39955112 - 39969968 on Build GRCh38
UniGene ID
Hs.655177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136493.2 NP_001129965.1 13 1621
NM_001287808.1 NP_001274737.1 11 1322
NM_001287809.1 NP_001274738.1 13 1465
NM_032793.4 NP_116182.2 13 1582
NR_109896.1 11 1385
XM_005271285.1 XP_005271342.1 13 1546
XM_011542312.2 XP_011540614.1 12 1465
GenBank mRNA AB073383.1 - 13 1546
AF289609.1 - 13 1538
AF370364.1 - 8 1842
AK074186.1 - 13 1487
AK075183.1 - 13 1549
AK093223.1 - 13 1583
AK291540.1 - 13 1544
AK297991.1 - 13 1429
AK301533.1 - 11 1289
AL834184.1 - 11 1339
AY358636.1 - 13 1564
BC006353.2 - 9 917
BC011587.1 - 13 1556 4646430
BC092414.1 - 13 1524 30387073

View Full Product Details