Gene Transcripts

Gene Symbol
DOCK7
Entrez Gene ID
85440
Gene Name
dedicator of cytokinesis 7
Gene Aliases
EIEE23, ZIR2
Chromosome Location
Chr. 1: 62454726 - 62688368 on Build GRCh38
UniGene ID
Hs.744927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271999.1 NP_001258928.1 24 2977
NM_001272000.1 NP_001258929.1 23 2884
NM_001272001.1 NP_001258930.1 23 2884
NM_033407.3 NP_212132.2 23 2884
XM_005271292.2 XP_005271349.1 24 2967
XM_011542326.2 XP_011540628.1 24 2967
XM_011542327.2 XP_011540629.1 24 2967
XM_011542328.2 XP_011540630.1 24 2967
XM_011542330.2 XP_011540632.1 24 2967
XM_017002639.1 XP_016858128.1 23 2874
XM_017002640.1 XP_016858129.1 24 2967
GenBank mRNA AB051558.1 - 4 387
AK125049.1 - 9 1029
DQ118679.1 - 20 2456
DQ118680.1 - 20 2456
DQ341187.1 - 24 2884

View Full Product Details