Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193416.2 | NP_001180345.1 | 11 | 1144 | |
NM_001193417.2 | NP_001180346.1 | 10 | 1096 | ||
NM_001356.4 | NP_001347.3 | 11 | 1998 | ||
NR_126093.1 | 11 | 1998 | |||
XM_011543892.2 | XP_011542194.1 | 11 | 1175 | ||
XM_017029313.1 | XP_016884802.1 | 11 | 3343 | ||
XM_017029314.1 | XP_016884803.1 | 11 | 3343 | ||
GenBank mRNA | AB208983.1 | - | 11 | 1096 | |
AB451220.1 | - | 11 | 1053 | ||
AB451343.1 | - | 11 | 1053 | ||
AF000982.1 | - | 11 | 1909 | ||
AF000983.1 | - | 11 | 1909 | ||
AF061337.1 | - | 11 | 1128 | ||
AK291153.1 | - | 11 | 1144 | ||
AK297159.1 | - | 11 | 1144 | ||
AK304661.1 | - | 10 | 1101 | ||
AK304689.1 | - | 10 | 1096 | ||
AY776161.1 | - | 3 | 204 | ||
BC011819.2 | - | 11 | 1123 | 3617040 | |
CB107740.1 | - | 5 | 412 | ||
U50553.1 | - | 11 | 1362 |