Gene Transcripts
- Gene Symbol
- MVB12B
- Entrez Gene ID
- 89853
- Gene Name
- multivesicular body subunit 12B
- Gene Aliases
- C9orf28, FAM125B
- Chromosome Location
- Chr. 9: 126326844 - 126507041 on Build GRCh38
- UniGene ID
- Hs.162659
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001011703.2 |
NP_001011703.1
|
2, 3 |
288 |
|
NM_033446.2 |
NP_258257.1
|
2, 3 |
288 |
|
XM_005252297.1 |
XP_005252354.1
|
2, 3 |
323 |
|
XM_017015275.1 |
XP_016870764.1
|
2, 3 |
288 |
|
XM_017015276.1 |
XP_016870765.1
|
2, 3 |
323 |
|
XM_017015277.1 |
XP_016870766.1
|
2, 3 |
288 |
|
XM_017015278.1 |
XP_016870767.1
|
2, 3 |
288 |
|
GenBank mRNA |
AK024432.1 |
- |
2, 3 |
283 |
|
AK293740.1 |
- |
2, 3 |
282 |
|
BC000122.1 |
- |
2, 3 |
263 |
|
BC028675.1 |
- |
2, 3 |
274 |
4083276 |
DA339911.1 |
- |
2, 3 |
288 |
|
View Full Product Details