Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033271.2 | NP_150374.2 | 5 | 1273 | |
XM_005268188.3 | XP_005268245.1 | 4 | 1441 | ||
GenBank mRNA | AF173358.1 | - | 1 | 1019 | |
AF353674.1 | - | 4 | 1137 | ||
BC006414.1 | - | 1 | 60 | ||
BC042525.1 | - | 5 | 1243 | 4824525 |