Gene Transcripts
- Gene Symbol
- TBL1Y
- Entrez Gene ID
- 90665
- Gene Name
- transducin (beta)-like 1, Y-linked
- Gene Aliases
- TBL1
- Chromosome Location
- Chr. Y: 6910686 - 7107154 on Build GRCh38
- UniGene ID
- Hs.664560
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033284.1 |
NP_150600.1
|
7, 8 |
843 |
|
NM_134258.1 |
NP_599020.1
|
6, 7 |
812 |
|
NM_134259.1 |
NP_599021.1
|
6, 7 |
748 |
|
XM_005262572.3 |
XP_005262629.1
|
1, 2 |
507 |
|
XM_017030086.1 |
XP_016885575.1
|
7, 8 |
832 |
|
XM_017030087.1 |
XP_016885576.1
|
7, 8 |
832 |
|
GenBank mRNA |
AF332220.1 |
- |
7, 8 |
843 |
|
AF332221.1 |
- |
6, 7 |
812 |
|
AF332222.1 |
- |
6, 7 |
748 |
|
AK298210.1 |
- |
6, 7 |
724 |
|
BC130471.1 |
- |
3, 4 |
326 |
40146502 |
BC130473.1 |
- |
3, 4 |
326 |
40146504 |
View Full Product Details