Gene Transcripts

Gene Symbol
CERS5
Entrez Gene ID
91012
Gene Name
ceramide synthase 5
Gene Aliases
LASS5, Trh4
Chromosome Location
Chr. 12: 50129306 - 50167533 on Build GRCh38
UniGene ID
Hs.270525
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281731.1 NP_001268660.1 8 1138
NM_147190.3 NP_671723.1 7 937
NR_104035.1 6 879
XM_005269220.2 XP_005269277.1 7 773
XM_006719687.3 XP_006719750.1 7 773
XM_011538975.2 XP_011537277.1 7 773
XM_017020202.1 XP_016875691.1 7 697
XM_017020203.1 XP_016875692.1 8 1061
XM_017020204.1 XP_016875693.1 7 773
XM_017020205.1 XP_016875694.1 7 626
GenBank mRNA AK026014.1 - 7 743
AK058033.1 - 5 1329
AK294521.1 - 6 653
AK298924.1 - 6 691
AK300937.1 - 8 913
BC032565.1 - 7 754 5580489
BC033558.1 - 6 666
U15426.1 - 801

View Full Product Details