Gene Transcripts
- Gene Symbol
- DDX60L
- Entrez Gene ID
- 91351
- Gene Name
- DEAD-box helicase 60-like
- Gene Aliases
- Chromosome Location
- Chr. 4: 168356735 - 168480514 on Build GRCh38
- UniGene ID
- Hs.535011
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012967.2 |
NP_001012985.2
|
28, 29 |
4065 |
|
NM_001291510.1 |
NP_001278439.1
|
28, 29 |
4068 |
|
XM_005263341.4 |
XP_005263398.2
|
28, 29 |
4051 |
|
XM_005263342.2 |
XP_005263399.2
|
28, 29 |
4068 |
|
XM_005263343.2 |
XP_005263400.2
|
28, 29 |
4034 |
|
XM_005263345.4 |
XP_005263402.2
|
28, 29 |
4047 |
|
XM_006714408.2 |
XP_006714471.1
|
29, 30 |
4118 |
|
XM_011532402.2 |
XP_011530704.1
|
29, 30 |
4341 |
|
XM_011532403.2 |
XP_011530705.1
|
29, 30 |
4176 |
|
XM_011532404.1 |
XP_011530706.1
|
27, 28 |
3972 |
|
XM_011532405.1 |
XP_011530707.1
|
27, 28 |
3969 |
|
XM_011532406.1 |
XP_011530708.1
|
28, 29 |
4068 |
|
XM_011532409.2 |
XP_011530711.1
|
16, 17 |
2108 |
|
XM_017008829.1 |
XP_016864318.1
|
27, 28 |
3938 |
|
XM_017008831.1 |
XP_016864320.1
|
17, 18 |
2370 |
|
GenBank mRNA |
AK023743.1 |
- |
7, 8 |
951 |
|
AL832950.1 |
- |
9, 10 |
1160 |
|
CR933601.1 |
- |
28, 29 |
4043 |
|
KR632542.1 |
- |
26, 27 |
3817 |
|
View Full Product Details