Gene Transcripts

Gene Symbol
RASL10B
Entrez Gene ID
91608
Gene Name
RAS like family 10 member B
Gene Aliases
RRP17, VTS58635
Chromosome Location
Chr. 17: 35731660 - 35743521 on Build GRCh38
UniGene ID
Hs.744994
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033315.3 NP_201572.1 4 1818
XM_017025300.1 XP_016880789.1 5 2712
XM_017025301.1 XP_016880790.1 3 2452
GenBank mRNA AK122652.1 - 4 1816
BC041133.1 - 4 1738 5762715
DA250459.1 - 1 415

View Full Product Details